Skip to Content
Merck
All Photos(1)

Key Documents

95314

Sigma-Aldrich

Hydrogen peroxide–Urea adduct

purum p.a., "rapid-soluble", tablet (1 g each)

Synonym(s):

Hydrogen peroxide–Urea adduct, Carbamide Per hydrate, Carbamide peroxide, Percarbamide, Urea hydrogen peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CO(NH2)2 · H2O2
CAS Number:
Molecular Weight:
94.07
Beilstein:
3680414
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

23.3 mmHg ( 30 °C)

grade

purum p.a.

form

tablet (1 g each)

quality

"rapid-soluble"

composition

active oxygen, 15-17%

reaction suitability

reagent type: oxidant

mp

90-93 °C (lit.)

functional group

amine

SMILES string

OO.NC(N)=O

InChI

1S/CH4N2O.H2O2/c2-1(3)4;1-2/h(H4,2,3,4);1-2H

InChI key

AQLJVWUFPCUVLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant which undergoes solid-state decomposition as a source of peroxide

Reactant used as an oxidant

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. O. Mahmoodi, et al.,
Synthetic Communications, 40, 3181-3185 (2010)
Donghua Alan Zhu et al.
Journal of pharmaceutical sciences, 100(8), 3529-3538 (2011-04-27)
Despite the fact that oxidation is the second most frequent degradation pathway for pharmaceuticals, means of evaluating the oxidative stability of pharmaceutical solids, especially effective stress testing, are still lacking. This paper describes a novel experimental method for peroxide-mediated oxidative
T. Michel, et al.,
J. Mol. Catal. A: Chem., 340, 9-14 (2011)
Karen Pintado-Palomino et al.
Journal of dentistry, 43(9), 1099-1105 (2015-07-15)
To evaluate the efficacy of experimental proposals of desensitizing agents during tooth bleaching. 140 participants without tooth sensitivity (TS) received 16% carbamide peroxide (14 days-04 h each) (T1) or 35% hydrogen peroxide (single session-45 min) (T2). Participants used concomitantly (10
Lidia Yileng Tay et al.
American journal of dentistry, 25(4), 199-204 (2012-10-23)
This parallel, double-blind randomized clinical trial evaluated the 2-year bleaching efficacy and sensitivity produced by at-home (AH) and in-office (IO) bleaching therapies. 60 participants with tooth color darker than C2, without restorations in the anterior dentition and older than 18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service