Skip to Content
Merck
All Photos(1)

Key Documents

93970

Sigma-Aldrich

Ucon HTF 14

range of application 18-260 °C

Synonym(s):

Poly(ethylene glycol-ran-propylene glycol) monobutyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

range of application measuring range

18-260 °C

density

1.04 g/mL at 20 °C

SMILES string

OCCO.CCCCO.CC(O)CO

InChI

1S/C4H10O.C3H8O2.C2H6O2/c1-2-3-4-5;1-3(5)2-4;3-1-2-4/h5H,2-4H2,1H3;3-5H,2H2,1H3;3-4H,1-2H2

InChI key

QMNOIORHZMRPLS-UHFFFAOYSA-N

Application

Ucon HTF 14 is a thermoseparating polymer (TSP) that can be used in aqueous two-phase extraction (ATPE) due to its decreasing solubility in its aqueous solution as the temperature rises.[1] It is also used in a polymer coating on solid-phase microextraction (SPME) fibers using the sol-gel technique to impart thermal resistance and stability.[1][2]
water soluble oil bath fluid

Legal Information

Ucon is a trademark of Union Carbide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sol-gel-based solid-phase microextraction and gas chromatography-mass spectrometry determination of dextromethorphan and dextrorphan in human plasma
Bagheri, Habib and Es-haghi, Ali and Rouini, Mohammad-Reza
Journal of Chromatography. B, Biomedical Sciences and Applications, 818(2), 147-157 (2005)
Wei Zhang et al.
Biomaterials, 32(11), 2894-2906 (2011-01-25)
The aim of this study was to exploit the possibility of combination of active targeting function of folic acid by folate receptor-mediated endocytosis and overcoming multidrug resistance (MDR) by Pluronic block copolymers to promote drug delivery to MDR tumor following
Juan Ramón Avendaño-Gómez et al.
Journal of colloid and interface science, 380(1), 75-82 (2012-06-02)
The aim of this works is to study an oil-in-water emulsion stabilized with a triblock copolymer Synperonic F127 which presents a double size distribution of oil droplets. The emulsions were studied experimentally by means of differential scanning calorimetry (DSC) and
Céline Durand-Gasselin et al.
Langmuir : the ACS journal of surfaces and colloids, 26(14), 12321-12329 (2010-06-10)
Two amino-terminated amphiphilic copolymers, M600 and M1000, with different ethylene oxide to propylene oxide EO/PO ratios, 1/9 and 19/3, respectively, were coupled by thioctic acid, which allows an excellent affinity with gold surface. Then, amphiphilic thermally responsive gold nanoparticles (AuNPs)
Hong Ding et al.
Nanoscale, 3(4), 1813-1822 (2011-03-03)
In this study QDs were encapsulated in carboxylated PluronicF127 (F127COOH) triblock polymeric micelles and conjugated with anti-mesothelin antibody for the purpose of alleviating potential toxicity, enhancing the stability and improving targeting efficiency of CdTe/ZnS quantum dots (QDs) in tumors. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service