Skip to Content
Merck
All Photos(1)

Documents

936405

Sigma-Aldrich

Fmoc-Val-Cit-PAB-OH

≥95%

Synonym(s):

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-valyl-N5-(aminocarbonyl)-N-[4-(hydroxymethyl)phenyl]-L-ornithinamide, L-Ornithinamide, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-valyl-N5-(aminocarbonyl)-N-[4-(hydroxymethyl)phenyl]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H39N5O6
CAS Number:
Molecular Weight:
601.69
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

Assay

≥95%

form

solid

color

white to off-white

storage temp.

2-8°C

SMILES string

OCC(C=C1)=CC=C1NC([C@H](CCCNC(N)=O)NC([C@H](C(C)C)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)=O)=O

InChI

InChI= 1S/C33H39N5O6/c1-20(2)29(38-33(43)44-19-27-25-10-5-3-8-23(25)24-9-4-6-11-26(24)27)31(41)37-28(12-7-17-35-32(34)42)30(40)36-22-15-13-21(18-39)14-16-22/h3-6,8-11,13-16,20,27-29,39H,7,12,17-19H2,1-2H3,(H,36,40)(H,37,41)(H,38,43)(H3,34,35,42)/t28-,29-/m0/s1

Application

This is a cathepsin cleavable linker used in the construction of antibody drug conjugates (ADC & Bioconjugation).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EGFR Targeted Cetuximab-Valine-Citrulline (vc)-Doxorubicin Immunoconjugates- Loaded Bovine Serum Albumin (BSA) Nanoparticles for Colorectal Tumor Therapy
Ye Z, et al.
International journal of nanomedicine, 26, 2443-2459 (2021)
Cryptophycin-55/52 based antibody-drug conjugates: Synthesis, efficacy, and mode of action studies
Lai Q, et al.
European Journal of Medicinal Chemistry, 199, 112364-112364 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service