Skip to Content
Merck
All Photos(1)

Documents

934372

Sigma-Aldrich

MC-Val-Cit-PAB-PNP

≥95%

Synonym(s):

L-Ornithinamide, N-[6-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-1-oxohexyl]-L-valyl-N5-(aminocarbonyl)-N-[4-[[[(4-nitrophenoxy)carbonyl]oxy]methyl]phenyl]-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H43N7O11
CAS Number:
Molecular Weight:
737.76
Beilstein:
10322169
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

Application

This is a bifunctional cathepsin cleavable linker used in the construction of antibody drug conjugates (ADC).

Features and Benefits

The maleimide functionality can be used for bioconjugation to thiols in monoclonal antibodies while the activated carbonate esters can be used for construction of carbamate linkages with amine containing payloads.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zixuan Ye et al.
International journal of nanomedicine, 16, 2443-2459 (2021-04-06)
Specific modifications to carriers to achieve targeted delivery of chemotherapeutics into malignant tissues are a critical point for efficient diagnosis and therapy. In this case, bovine serum albumin (BSA) was conjugated with cetuximab-valine-citrulline (vc)-doxorubicin (DOX) to target epidermal growth factor
Qinhuai Lai et al.
European journal of medicinal chemistry, 199, 112364-112364 (2020-05-14)
Cryptophycin-52 (CR52), a tubulin inhibitor, exhibits promising antitumor activity in vitro (picomolar level) and in mouse xenograft models. However, the narrow therapeutic window in clinical trials limits its further development. Antibody-drug conjugate (ADC), formed by coupling cytotoxic compound (payload) to an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service