Skip to Content
Merck
All Photos(1)

Documents

934356

Sigma-Aldrich

Val-cit-PAB-OH

≥95%

Synonym(s):

L-Valyl-N5-(aminocarbonyl)-N-[4-(hydroxymethylphenyl]-L-ornithinamide, L-Valyl-N5-(aminocarbonyl)-N-[4-(hydroxymethyl)phenyl]-L-ornithinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H29N5O4
CAS Number:
Molecular Weight:
379.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

This is a cathepsin cleavable linker used in the construction of antibody drug conjugates (ADC).

Cathepsin cleaveable linkers

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinhuai Lai et al.
European journal of medicinal chemistry, 199, 112364-112364 (2020-05-14)
Cryptophycin-52 (CR52), a tubulin inhibitor, exhibits promising antitumor activity in vitro (picomolar level) and in mouse xenograft models. However, the narrow therapeutic window in clinical trials limits its further development. Antibody-drug conjugate (ADC), formed by coupling cytotoxic compound (payload) to an
Zixuan Ye et al.
International journal of nanomedicine, 16, 2443-2459 (2021-04-06)
Specific modifications to carriers to achieve targeted delivery of chemotherapeutics into malignant tissues are a critical point for efficient diagnosis and therapy. In this case, bovine serum albumin (BSA) was conjugated with cetuximab-valine-citrulline (vc)-doxorubicin (DOX) to target epidermal growth factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service