Skip to Content
Merck
All Photos(1)

Key Documents

933112

Sigma-Aldrich

SEM Sample Holder

EMS Orbital Specimen Holder, Pin Mount

Synonym(s):

Orbital Mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35
Pricing and availability is not currently available.

Looking for similar products? Visit Product Comparison Guide

General description

EMS Techware

Application

Take the new EMS Orbital Specimen Holder for a spin and raise your SEM images to a whole new level. Experience flexibility and control superior to any other specimen holder. This unique holder is a tool no SEM laboratory should be without.
The EMS Orbital Specimen Holder′s clever design allows you to effortlessly rotate the specimen completely around an axis. The pot is coated to reduce noise, producing a black background behind the subject so every detail stands out, allowing the capture of stunning, clear images.

Features and Benefits

The EMS Orbital Specimen Holder′s clever design allows you to effortlessly rotate the specimen 60° around the axis perpendicular to the electron beam. The position and hold of the sample can be controlled with the simple turn of a screw.
  • Rotate 360° around the axis perpendicular to the electron beam
  • Pot is detached, fully re-positionable below the pivoted mount
  • Control and hold the position of the sample with the simple turn of a screw
  • Overall Dimensions: Pin Mount - 1.25" (L) x 1" (W) x 1.5" (H)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service