Skip to Content
Merck
All Photos(1)

Key Documents

927090

Sigma-Aldrich

Ni(COD)(CPDO-(OMe)Ph)

≥95%

Synonym(s):

(1,5-cyclooctadiene)(2,3,4,5-tetrakis(4-methoxyphenyl)cyclopenta-2,4-dien-1-one)nickel(0)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$250.00
500 MG
$783.00

$250.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 MG
$250.00
500 MG
$783.00

About This Item

Empirical Formula (Hill Notation):
C41H40NiO5
Molecular Weight:
671.45
UNSPSC Code:
12352103
NACRES:
NA.21

$250.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥95%

form

powder or chunks

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

storage temp.

2-8°C

Application

Ni(COD)(CPDO-(OMe)Ph) is a bench-stable 18-electron nickel–olefin complex that can be used for a variety of reactions.

Reference

Structurally Diverse Bench-Stable Nickel(0) Pre-Catalysts: A Practical Toolkit for In Situ Ligation Protocols

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Engle lab pioneers efficient catalytic methods, leveraging substrate directivity for streamlined organic synthesis and selective product formation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service