Skip to Content
Merck
All Photos(1)

Key Documents

931888

Sigma-Aldrich

Hydrophobic MXene for composites

Synonym(s):

Hydrophobic MXenes with alkylammonium surface modification

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H33NH3-Ti3C2Tx
UNSPSC Code:
12352200
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.9% trace metals basis

Quality Level

impurities

≤1000 ppm (trace metals)

color

dark gray to black

SMILES string

CCCCCCCCCCCCCCCC[NH3+].[Ti3C2Tx]

Application

This product is a Ti3C2 MXene material that has been modified by addition of a hydrophobic cationic ammonium salt onto the surface. Intercalation of hydrophobic cations into MXene layers improves the dispersibility in non-aqueous solvents. These materials give you a MXene material with stability of and higher versitility as more solvent systems are unlocked.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Gastrointestinal tract,Liver,Immune system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering the Interlayer Spacing by Pre-Intercalation for High Performance Supercapacitor MXene Electrodes in Room Temperature Ionic Liquid
Liang, K.; et al.
Advanced Functional Materials , 31, 2104007-2104007 (2021)
Alkylammonium Cation Intercalation into Ti3C2 (MXene): Effects on Properties and Ion-Exchange Capacity Estimation
Ghidiu, M., et al.
Chemistry of Materials, 29, 1099-1099 (2017)
Min Xu et al.
ACS nano, 12(4), 3733-3740 (2018-03-16)
Two-dimensional (2D) Ti3C2 MXene has attracted great attention in electrochemical energy storage devices (supercapacitors and lithium-ion and sodium-ion batteries) due to its excellent electrical conductivity as well as high volumetric capacity. Nevertheless, a previous study showed that multivalent Mg2+ ions
Dispersion and Stabilization of Alkylated 2D MXene in Nonpolar Solvents and Their Pseudocapacitive Behavior
Carey, M., et al.
Cell reports. Physical science, 1, 100042-100042 (2020)

Articles

Discover how MXenes' superior conductivity, strength, and stability are revolutionizing energy. Explore their potential for next-gen batteries and hydrogen evolution.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service