Skip to Content
Merck
All Photos(1)

Key Documents

931845

Sigma-Aldrich

GenOx-PJ

Synonym(s):

3-Oxo-2-tosyl-2,3-dihydro-1H-1λ3-benzo[d][1,2]iodazol-1-yl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14INO5S
CAS Number:
Molecular Weight:
459.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

form

powder

Quality Level

mp

>300 °C

functional group

iodo

storage temp.

−20°C

Related Categories

Application

GenOx-PJ is an iodine(III) based oxidant developed by Genentech as part of a robust method of forming C-O bonds through direct coupling of carboxylic acids with alcohols. When used with a Ru(II) photocatalyst, GenOx-PJ is active in the decarboxylative coupling of a broad range of carboxylic acids--including primary, secondary, tertiary, as well as peptides--with alcohols. By this method, GenOx-PJ enables a direct approach in utilizing simple, readily available substrates to perform this type of coupling, and providing many synthetic applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Platform for Decarboxylative Couplings via Photoredox Catalysis: Direct Access to Carbocations from Carboxylic Acids for Carbon?Oxygen Bond Formation
Li P, et al.
ACS Catalysis, 11(17), 10997?11004-10997?11004 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service