Skip to Content
Merck
All Photos(1)

Key Documents

925519

Sigma-Aldrich

BpyCAM•HCl

Synonym(s):

2,2′-Bipyridine-6-carboxamidine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$149.00
500 MG
$483.00

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$149.00
500 MG
$483.00

About This Item

Empirical Formula (Hill Notation):
C11H11ClN4
CAS Number:
Molecular Weight:
234.68
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>215 °C

storage temp.

−20°C

SMILES string

NC(C1=CC=CC(C2=CC=CC=N2)=N1)=N.Cl

InChI

1S/C11H10N4.ClH/c12-11(13)10-6-3-5-9(15-10)8-4-1-2-7-14-8;/h1-7H,(H3,12,13);1H

InChI key

DWSUJJYYHWLZMW-UHFFFAOYSA-N

Application

BpyCAM•HCl is a bipyridine carboxamidine ligand that enables nickel-catalyzed cross-electrophile couplings of diverse aryl and alkyl halides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service