Skip to Content
Merck
All Photos(2)

Key Documents

902039

Sigma-Aldrich

4-Methoxypicolinimidamide hydrochloride

≥95%

Synonym(s):

Weix 2-pyridylcarboxamidine ligand for nickel catalysis

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$223.00
1 G
$449.00

$223.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$223.00
1 G
$449.00

About This Item

Empirical Formula (Hill Notation):
C7H10ClN3O
CAS Number:
Molecular Weight:
187.63
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$223.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder or solid

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

mp

174-176 °C

SMILES string

Cl.N\C(=N/[H])\c1nccc(c1)OC

InChI

1S/C7H9N3O.ClH/c1-11-5-2-3-10-6(4-5)7(8)9;/h2-4H,1H3,(H3,8,9);1H

InChI key

JOUDWIBACXLPIO-UHFFFAOYSA-N

Application

This pyridyl carboxamidine ligand has been demonstrated by Daniel Weix′s lab to enable nickel-catalyzed cross-coupling of diverse basic nitrogen heterocycles with primary and secondary alkyl halides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric C Hansen et al.
The Journal of organic chemistry, 82(14), 7085-7092 (2017-07-07)
Despite their importance, the synthesis of alkylated heterocycles from the cross-coupling of Lewis basic nitrogen heteroaryl halides with alkyl halides remains a challenge. We report here a general solution to this challenge enabled by a new collection of ligands based
Robert J Perkins et al.
Organic letters, 19(14), 3755-3758 (2017-07-14)
A constant-current electrochemical method for reducing catalytic nickel complexes in sp2-sp3 cross-electrophile coupling reactions has been developed. The electrochemical reduction provides reliable nickel catalyst activation and turnover and offers a tunable parameter for reaction optimization, in contrast to more standard
Eric C Hansen et al.
Nature chemistry, 8(12), 1126-1130 (2016-11-23)
Ligands are essential for controlling the reactivity and selectivity of reactions catalysed by transition metals. Access to large phosphine ligand libraries has become an essential tool for the application of metal-catalysed reactions industrially, but these existing libraries are not well

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service