Skip to Content
Merck
All Photos(1)

Key Documents

917974

Sigma-Aldrich

BocA1V2PF2

Synonym(s):

(S)-2-((S)-1-((S)-2-((S)-2-((tert-Butoxycarbonyl)amino)propanamido)-2-cyclohexylacetyl)pyrrolidine-2-carboxamido)-3-(4-fluorophenyl)propanoic acid, IAP E3 ligase lead for protein degrader research, SNIPER building block

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$615.00

$615.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$615.00

About This Item

Empirical Formula (Hill Notation):
C30H43FN4O7
Molecular Weight:
590.68
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.22

$615.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

ligand

BocA1V2PF2

Quality Level

form

powder

reaction suitability

reagent type: ligand

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

C[C@H](NC(OC(C)(C)C)=O)C(N[C@H](C(N1CCC[C@H]1C(N[C@H](C(O)=O)CC2=CC=C(C=C2)F)=O)=O)C3CCCCC3)=O

InChI

1S/C30H43FN4O7/c1-18(32-29(41)42-30(2,3)4)25(36)34-24(20-9-6-5-7-10-20)27(38)35-16-8-11-23(35)26(37)33-22(28(39)40)17-19-12-14-21(31)15-13-19/h12-15,18,20,22-24H,5-11,16-17H2,1-4H3,(H,32,41)(H,33,37)(H,34,36)(H,39,40)/t18-,22-,23-,24-/m0/s1

InChI key

LNJCHGKSSPMNRM-IMNFJDCFSA-N

Application

BocA1V2PF2 is an in silico-derived inhibitor of apoptosis protein (IAP)-recruiting ligand for targeted protein degradation and SNIPER (specific and non-genetic IAP-dependent protein erasers) development, launched in partnership with ComInnex. Learn more about the novel IAP ligands generated through virtual screening of AVP mimetics in our Technology Spotlights. A C-terminal variant of BocA1V2PF2 is also available as A1V2PF2-NHEt (916714).

BocA1V2PF2 conjugates are also available for degrader synthesis. Browse our full synthesis offering here for streamlining
SNIPER and PROTAC® degrader libraries: Degrader Building Blocks

917443BocA1V2PF2-NHC6-NH2
917699 BocA1V2PF2-NHC10-NH2
917958 BocA1V2PF2-NHPEG1-NH2
916692 BocA1V2PF2-NHPEG3-NH2

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobumichi Ohoka et al.
The Journal of biological chemistry, 292(11), 4556-4570 (2017-02-06)
Many diseases, especially cancers, result from aberrant or overexpression of pathogenic proteins. Specific inhibitors against these proteins have shown remarkable therapeutic effects, but these are limited mainly to enzymes. An alternative approach that may have utility in drug development relies
Tasuku Ishida et al.
SLAS discovery : advancing life sciences R & D, 26(4), 484-502 (2020-11-05)
Bifunctional degrader molecules, also called proteolysis-targeting chimeras (PROTACs), are a new modality of chemical tools and potential therapeutics to understand and treat human disease. A required PROTAC component is a ligand binding to an E3 ubiquitin ligase, which is then joined to another ligand binding to a protein to
Mikihiko Naito et al.
Drug discovery today. Technologies, 31, 35-42 (2019-06-16)
The induction of protein degradation by chimeric small molecules represented by proteolysis-targeting chimeras (PROTACs) is an emerging approach for novel drug development. We have developed a series of chimeric molecules termed specific and non-genetic inhibitor of apoptosis protein (IAP)-dependent protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service