917699
BocA1V2PF2-NHC10-NH2
≥85%
Synonym(s):
tert-Butyl ((S)-1-(((S)-2-((S)-2-(((S)-1-((10-Aminodecyl)amino)-3-(4-fluorophenyl)-1-oxopropan-2-yl)carbamoyl)pyrrolidin-1-yl)-1-cyclohexyl-2-oxoethyl)amino)-1-oxopropan-2-yl)carbamate, AVP conjugate for IAP-mediated protein degrader development, SNIPER building block
Select a Size
Select a Size
About This Item
Recommended Products
ligand
BocA1V2PF2
Quality Level
Assay
≥85%
form
powder
reaction suitability
reactivity: carboxyl reactive
reagent type: ligand-linker conjugate
functional group
amine
storage temp.
2-8°C
SMILES string
C[C@H](NC(OC(C)(C)C)=O)C(N[C@H](C(N1CCC[C@H]1C(N[C@H](C(NCCCCCCCCCCN)=O)CC2=CC=C(C=C2)F)=O)=O)C3CCCCC3)=O
InChI
1S/C40H65FN6O6/c1-28(44-39(52)53-40(2,3)4)35(48)46-34(30-17-12-11-13-18-30)38(51)47-26-16-19-33(47)37(50)45-32(27-29-20-22-31(41)23-21-29)36(49)43-25-15-10-8-6-5-7-9-14-24-42/h20-23,28,30,32-34H,5-19,24-27,42H2,1-4H3,(H,43,49)(H,44,52)(H,45,50)(H,46,48)/t
InChI key
AFZPWVXOGIXQBS-MTZHBSBLSA-N
Related Categories
Application
Building blocks in this series:
917974 BocA1V2PF2
917443 BocA1V2PF2-NHC6-NH2
917699 BocA1V2PF2-NHC10-NH2
916951 BocA1V2PF1-NHPEG1-NH2
917214 BocA1V2PF1-NHPEG3-NH2
Technology Spotlight: Degrader Building Blocks with Inhibitor of Apoptosis Protein (IAP) In Silico-Derived Ligands
Other Notes
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Targeted protein degradation reduces disease-relevant proteins in cells using small molecules, hijacking endogenous proteolysis systems.
Plate of 80 ligands against E3 ligase IAP designed by ComInnex; allows creation of bifunctional targeted protein degraders or molecular glues.
Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service