Skip to Content
Merck
All Photos(1)

Key Documents

916153

Sigma-Aldrich

Lithium modified silica nanoparticles

pore size non-porous, 400 nm particle size

Synonym(s):

Battery electrolytes, Nanosalt, Silica functionalized li-ion particles

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$316.00

$316.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$316.00

About This Item

Linear Formula:
SiO2O(CH2)3SO3-Li+
CAS Number:
MDL number:
UNSPSC Code:
12352309
NACRES:
NA.23

$316.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

particle size

400 nm

pore size

non-porous

application(s)

battery manufacturing

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Monodisperse nonporous silica nanoparticles with Lithium ion functionalization have been used in electrolytes and nanosalts[1][2][3]. We offer highly monodisperse nonporous silica particles with lithium tertbutoxide, with sizes varying from 200 nm to 600 nm.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supercapacitive properties of activated carbon electrode in electrolyte solution with a lithium-modified silica nanosalt.
Jung H W, et al.
Current Applied Physics, 2015, 567-570 null
Gel electrolytes based on lithium modified silica nano-particles.
Sun J, et al.
Electrochimica Acta, 52, 7083-7090 (2007)
Lithium modified silica as electrolyte additive for lithium secondary batteries.
Latifatu M, et al.
Solid State Ionics, 319, 7-12 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service