Skip to Content
Merck
All Photos(1)

Key Documents

913650

Sigma-Aldrich

Polyoxyethylene bis(azide) solution

average Mn 450, 0.1 g/mL in THF

Synonym(s):

O,O′-Bis(2-azidoethyl)polyethylene glycol, PEG-diazide

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$174.00

$174.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$174.00

About This Item

Linear Formula:
N3(C2H4O)nC2H4N3
UNSPSC Code:
12352125
NACRES:
NA.23

$174.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

mol wt

PEG ~400 Da
average Mn 450

concentration

0.1 g/mL in THF

color

pale yellow

storage temp.

2-8°C

Application

PEG for cross-linking and used in "click" chemistry reactions. PEGylated materials have found broad use in drug delivery systems, virology, and immunology, as the incorporation of PEG improves pharmacological properties such as increased water solubility, enhanced resistance to degradation (protein hydrolysis), increased circulation half-life, and reduced antigenicity. Polymer for preparing enzyme conjugates soluble in organic solvents; promising drug carrier.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

0.5 °F

Flash Point(C)

-17.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving the tensile strength of carbon nanotube yarn via one-step double [2+1] cycloadditions.
H Kim, et al.
Korean Journal of Chemical Engineering, 33(1), 299-304 (2016)
Shao Qiong Liu et al.
Biomaterials, 30(8), 1453-1461 (2008-12-23)
In this study, biodegradable PEG-peptide hydrogels have been synthesized using Click chemistry. A series of Arg-Gly-Asp (RGD) containing peptides were prepared via a solid phase synthesis approach, which were further functionalized with azide to yield peptide azide or peptide diazide.
Tilman Läppchen et al.
European journal of medicinal chemistry, 89, 279-295 (2014-12-03)
Calixarene 0118 is a potent anti-angiogenic agent that effectively inhibited tumor growth in preclinical studies, and is currently being evaluated in a phase I clinical trial. We have designed two close mimetics of calixarene 0118 containing a terminal alkynyl-functional group

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service