Skip to Content
Merck
All Photos(1)

Key Documents

756598

Sigma-Aldrich

Poly(ethylene glycol) bisazide

average Mn 1,100, azide

Synonym(s):

Polyoxyethylene bis(azide), Clickable PEG, PEG azide, Polyethylene glycol, O,O′-Bis(2-azidoethyl)polyethylene glycol, PEG-diazide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$466.00

$466.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$466.00

About This Item

Linear Formula:
N3CH2CH2(OCH2CH2)nN3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$466.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) bisazide, average Mn 1,100

form

solid or liquid

mol wt

average Mn 1,100

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

transition temp

Tm 29-35 °C

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service