Skip to Content
Merck
All Photos(2)

Key Documents

911550

Sigma-Aldrich

Bis-MPA-RAFT dendrimer

trimethylol propane core, generation 1

Synonym(s):

CTA functionalized dendrimer, Multi-arm RAFT

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$615.00

$615.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$615.00

About This Item

Empirical Formula (Hill Notation):
C99H104N6O18S12
Molecular Weight:
2050.69
UNSPSC Code:
12162002
NACRES:
NA.23

$615.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

mol wt

generation 1

no. Surface Groups

6

color

orange to red

storage temp.

−20°C

General description

  • Polymer architecture: Dendrimer
  • Generation: 1
  • End Group Functionality: RAFT
  • No. Surface Groups: 6
  • Calculated Mol. Wt.: 2050.7 g/mol
  • bis-MPA = 2,2-Bis(hydroxymethyl)propionic acid

Application

Bis-MPA dendritic molecules are based on 2,2-bismethylolpropionicacid (bis-MPA) building blocks. The hydrolysable linkers presented in this type of polymers allow their degradation into byproducts that can be reabsorbed by the body or into excretable non-toxic small molecules. Therefore, this group of dendritic polymers is biocompatible and biodegradable, making them suitable for biomedical applications.
The corona of this product is functionalized with RAFT agents, which allow direct controlled polymerization from the dendrimer surface. It can be readily used to construct multi-arm polymer structure using suitable acrylic monomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications
Garcia-Gallego, S. et al.
Progress in Polymer Science, 48, 85-110 (2015)
Patrik Stenström et al.
Molecules (Basel, Switzerland), 23(8) (2018-08-17)
Herein, we present the first evaluation of cationic dendrimers based on 2,2-bis(methylol)propionic acid (bis-MPA) as nonviral vectors for transfection of short interfering RNA (siRNA) in cell cultures. The study encompassed dendrimers of generation one to four (G1⁻G4), modified to bear
Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Articles

Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service