Skip to Content
Merck
All Photos(2)

Key Documents

900161

Sigma-Aldrich

4-((((2-Carboxyethyl)thio)carbonothioyl)thio)-4-cyanopentanoic acid

95%

Synonym(s):

4-[[[(2-Carboxyethyl)thio]thioxomethyl]thio]-4-cyanopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO4S3
CAS Number:
Molecular Weight:
307.41
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

powder

mp

101-106 °C

SMILES string

S(C(CCC(=O)O)(C)C#N)C(=S)SCCC(=O)O

InChI

1S/C10H13NO4S3/c1-10(6-11,4-2-7(12)13)18-9(16)17-5-3-8(14)15/h2-5H2,1H3,(H,12,13)(H,14,15)

InChI key

CTLAWKZAJCYIFX-UHFFFAOYSA-N

Application

A stable, trithiocarbonate chain transfer agent (CTA), also referred to as a RAFT agent, which can control chain growth in free radical polymerization producing polymers with well-defined molecular weights and low polydispersities.

4-((((2-Carboxyethyl)thio)carbonothioyl)thio)-4-cyanopentanoic acid may be used to control homo- and co-polymerization of acrylic acids, acrylates, acrylamides, styrenes, methacrylates, and methacrylamides. It is soluble in aqueous and organic media (amphiphilic).

Other Notes

Sold for research purposes only: See sigma-aldrich.com/raftlicense. Patents: WO98/01478, WO99/311444.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Low-cost, high-purity RAFT Agents are vital for industrial RAFT polymerization. We supply effective agents for this technology.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service