Skip to Content
Merck
All Photos(2)

Key Documents

911348

Sigma-Aldrich

Polyester bis-MPA dendron

generation 2, no. Surface Groups 4

Synonym(s):

Bis-MPA dendron, PFd-G1-Biotin-COOH

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$420.00

$420.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$420.00

About This Item

Empirical Formula (Hill Notation):
C49H72N2O28S
Molecular Weight:
1169.16
UNSPSC Code:
12162002
NACRES:
NA.23

$420.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

storage temp.

−20°C

General description

  • Polymer architecture: Dendron
  • Generation: 2
  • Core functional group: Biotin
  • End Group Functionality: Carboxylic acid
  • No. Surface Groups: 4
  • Calculated Mol. Wt.: 1169.16 g/mol
  • Bis-MPA = 2,2-Bis(hydroxymethyl)propionic acid

Application

Bis-MPA dendritic molecules are based on 2,2-bismethylolpropionicacid (bis-MPA) building blocks. The hydrolysable linkers presented in this type of polymers allows their degradation into byproducts that can be reabsorbed by the body or into excretable non-toxic small molecules. Therefore, this group of dendritic polymers are biocompatible and biodegradable, making them suitable for biomedical applications. The carboxylic acid-functionalized corona and biotin-functionalized core of this product can be readily used in bioconjugation reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications.
Garcia-Gallego S, et al.
Progress in Polymer Science, 48, 85-110 (2015)
Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications.
Garcia-Gallego S, et al
Progress in Polymer Science, 48, 85-110 (2015)
Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Articles

Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service