Skip to Content
Merck
All Photos(3)

Key Documents

910961

Sigma-Aldrich

di-n-butylammonium lead(II) tetraiodide

greener alternative

n = 1

Synonym(s):

(BA)2PbI4, 2D perovskite, di-n-butylammonium tetraiodoplumbate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$421.00
2.5 G
$842.00

$421.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$421.00
2.5 G
$842.00

About This Item

Linear Formula:
(C4H9NH3)2PbI4
CAS Number:
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

$421.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[Pb](I)(I)(I)I.[N+H3]CCCC.[N+H3]CCCC

InChI key

FMQPTTJYZPGGNR-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for "Energy efficiency".

Application

Di-n-butylammonium lead(II) tetra iodide is used as a precursor material in the fabrication of lead halide perovskite solar cells. Perovskite solar cells based on DBAPbI4 have shown high power conversion efficiencies and have attracted significant attention as a low-cost and efficient alternative to traditional silicon-based solar cells. DBAPbI4-based perovskite materials find application in thin-film electronics such as transistors and light-emitting diodes (LEDs).
Organic-inorganic hybrid perovskittes based semiconducting materials exhibit excellent properties such as long charge carrier lifetime and diffusion length, and high photolumiscence quantum yield. These properties render the perovskite materials useful for applications in light emitting diodes (LEDs), lasers and photodetectors. The bis[n-butylammonium] tetraiodopumbate is 2D-layered perovskite and finds applications as violet emitter in LEDs.[1][2][3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruddlesden?Popper Hybrid Lead Iodide Perovskite 2D Homologous Semiconductors
Chemistry of Materials, 28, 2852-2867 (2016)
Hsinhan Tsai et al.
Nature, 536(7616), 312-316 (2016-07-08)
Three-dimensional organic-inorganic perovskites have emerged as one of the most promising thin-film solar cell materials owing to their remarkable photophysical properties, which have led to power conversion efficiencies exceeding 20 per cent, with the prospect of further improvements towards the
Duyen H Cao et al.
Journal of the American Chemical Society, 137(24), 7843-7850 (2015-05-29)
We report on the fabrication and properties of the semiconducting 2D (CH3(CH2)3NH3)2(CH3NH3)(n-1)Pb(n)I(3n+1) (n = 1, 2, 3, and 4) perovskite thin films. The band gaps of the series decrease with increasing n values, from 2.24 eV (CH3(CH2)3NH3)2PbI4 (n = 1)

Articles

A brief tutorial on alternative energy materials for advanced batteries and fuel cells, as well as high-purity inorganics, conducting polymers, and electrolytes.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service