Skip to Content
Merck
All Photos(1)

Key Documents

91043

Sigma-Aldrich

L-Glutamic acid 5-tert-butyl 1-methyl ester hydrochloride

≥95.0% (TLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$185.00

$185.00


Estimated to ship onMarch 22, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$185.00

About This Item

Linear Formula:
(CH3)3COOCCH2CH2CH(NH2)COOCH3
CAS Number:
Molecular Weight:
253.72
Beilstein:
3572140
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$185.00


Estimated to ship onMarch 22, 2025


Request a Bulk Order

Quality Level

Assay

≥95.0% (TLC)

form

flakes

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cl[H].COC(=O)[C@@H](N)CCC(=O)OC(C)(C)C

InChI

1S/C10H19NO4.ClH/c1-10(2,3)15-8(12)6-5-7(11)9(13)14-4;/h7H,5-6,11H2,1-4H3;1H/t7-;/m0./s1

InChI key

YIFPACFSZQWAQF-FJXQXJEOSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service