Skip to Content
Merck
All Photos(1)

Key Documents

908339

Sigma-Aldrich

L-Methionine-13C5

≥99 atom % 13C, ≥98% (CP)

Synonym(s):

L-Methionine-13C5

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,360.00

$1,360.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$1,360.00

About This Item

Empirical Formula (Hill Notation):
13C5H11NO2S
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.12

$1,360.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥98% (CP)

form

solid

mass shift

M+5

SMILES string

O[13C]([13C@@H](N)[13CH2][13CH2]S[13CH3])=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i1+1,2+1,3+1,4+1,5+1

InChI key

FFEARJCKVFRZRR-JRGPAWSWSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service