Skip to Content
Merck
All Photos(1)

Key Documents

90625

Sigma-Aldrich

1,3,5-Triazine-2,4,6-trithiol trisodium salt solution

~15% in H2O, light yellow

Synonym(s):

Trimercapto-s-triazine trisodium salt, Trisodium 1,3,5-triazine-2,4,6-trithiolate solution, Trithiocyanuric acid trisodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$97.90

$97.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$97.90

About This Item

Empirical Formula (Hill Notation):
C3N3Na3S3
CAS Number:
Molecular Weight:
243.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$97.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solution

concentration

~15% in H2O

color

light yellow

SMILES string

[Na]Sc1nc(S[Na])nc(S[Na])n1

InChI

1S/C3H3N3S3.3Na/c7-1-4-2(8)6-3(9)5-1;;;/h(H3,4,5,6,7,8,9);;;/q;3*+1/p-3

InChI key

NYVOYAFUSJONHU-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohd Zulkefeli et al.
Inorganic chemistry, 48(19), 9567-9578 (2009-09-12)
A new supramolecular complex, {(Zn(4)L(4))(3)-(TCA(3-))(4)}(12+), was designed and synthesized by the 3:4 self-assembly of a linear tetrakis(Zn(2+)-cyclen) complex (Zn(4)L(4))(8+) and trianionic trithiocyanurate (TCA(3-)) in aqueous solution (cyclen = 1,4,7,10-tetraazacyclododecane). The {(Zn(4)L(4))(3)-(TCA(3-))(4)}(12+) complex, which should have a trigonal prism configuration, was
F J Koschier et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 21(4), 495-498 (1983-08-01)
Trithiocyanuric acid (TCY) is a rubber curative of low oral and dermal toxicity and with a low potential for eye and skin irritation. This study determined the effects of TCY on Sprague-Dawley rats at levels of 0.0, 625, 2500 and
Shin Aoki
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 122(12), 1095-1108 (2003-01-04)
We report novel supramolecular polyhedrons formed by three-dimensional self-assembly of multinuclear zinc(II)-12-membered tetraamine (= cyclen = 1,4,7,10-tetraazacyclododecane) complexes with potentially multidentate ligands such as cyanuric acid (CA) and trithiocyanuric acid (TCA) in aqueous solution. Two new supermolecular frameworks were isolated
Shin Aoki et al.
Chemistry, an Asian journal, 7(5), 944-956 (2012-02-09)
We have previously reported that the trimeric Zn(2+)-cyclen complex (tris(Zn(2+)-cyclen), [Zn(3)L(1)](6+)) and the trianion of trithiocyanuric acid (TCA(3-)) assembled in a 4:4 ratio to form a cuboctahedral supramolecular cage, [(Zn(3)L(1))(4)(TCA(3-))(4)](12+) (hereafter referred to as a Zn-cage), in neutral aqueous solution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service