Skip to Content
Merck
All Photos(1)

Key Documents

904899

Sigma-Aldrich

Spiro[9H-fluorene-9,9′-[9H]xanthene]-2,7-diamine

Synonym(s):

N,N,N′,N′-tetrakis(4-methoxyphenyl)spiro[fluorene-9,9′-xanthene]-2,7-diamine, X59

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$428.00

$428.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$428.00

About This Item

Empirical Formula (Hill Notation):
C53H42N2O5
CAS Number:
Molecular Weight:
786.91
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.23

$428.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

description

Band gap: Eg = 3.05 eV (lit)
Hole Mobility: 5.5 x 10-5 cm2/Vs (lit)

Assay

≥98%

form

powder

color

yellow

conductivity

1.9 x 10-4 S/cm (lit)

Orbital energy

HOMO -5.15 eV 
LUMO -2.10 eV 

SMILES string

N(c%10ccc(cc%10)OC)(c9ccc(cc9)OC)c1cc2c(cc1)c3c(cc(cc3)N(c8ccc(cc8)OC)c7ccc(cc7)OC)C42c5c(cccc5)Oc6c4cccc6

InChI key

PDGJIZDXBRVKBB-UHFFFAOYSA-N

General description

Spiro[9H-fluorene-9,9′-[9H]xanthene]-2,7-diamine (X59) is a hole transporting material (HTM), which has a spiro[fluorene-9,9′-xanthene] as a core component. It can be synthesized by Buchwald-Hartwig reaction. It shows a power conversion efficiency (PCE) of 19.8%.[1][2]

Application

X59 can be used in the formation of hole transporting layer (HTL) for the fabrication of polymeric solar cells (PSCs) and perovskite solar cells.[1][2][3][4]
X59 is a new hole transporting material (HTM) with spiro[fluorene-9,9′-xanthene] as the core moiety. An impressive power conversion efficiency (PCE) of 19.8% was achieved by using X59 as HTM in perovskite solar cell, which can compete with the record PCE by using the state-of-the-art-HTM Spiro-OMeTAD.[1][2] The X59-based devices show negligible hysteresis and reasonable stability in dark and dry conditions at room temperature for over five weeks.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strategy to modulate the pi-bridged units in bis (4-methoxyphenyl) amine-based hole-transporting materials for improvement of perovskite solar cell performance.
Liu H and Liu X
Journal of Material Chemistry C, 6(25), 6816-6822 (2018)
Rational design of bis(4-methoxyphenyl)amine-based molecules with different p-bridges as hole-transporting materials for efficient perovskite solar cells
Liu X, et al.
Dyes and Pigments, 139, 283-291 (2017)
Highly efficient and stable planar CsPbI2Br perovskite solar cell with a new sensitive-dopant-free hole transport layer obtained via an effective surface passivation.
Yang S, et al.
Solar Energy Materials and Solar Cells, 201(25), 110052-110052 (2019)
Facile synthesized organic hole transporting material for perovskite solar cell with efficiency of 19.8%
Bi Dongqin,et al.
Nano Energy, 23, 138-144 (2016)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service