Skip to Content
Merck
All Photos(1)

Key Documents

902012

Sigma-Aldrich

Molybdenum disulfide suspension

greener alternative

avg. no. of layers, < 3

Synonym(s):

2D-Moly sulfide dispersion, 2D-Molybdenum disulfide dipsersion with nonionic surfactant, 2D-Molybdenum disulfide dispersion, Moly sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$485.00

$485.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$485.00

About This Item

Linear Formula:
MoS2
UNSPSC Code:
12352302
NACRES:
NA.23

$485.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

50-1000 nm
thickness < 3 layers

form

liquid

feature

avg. no. of layers< 3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

5 mg/mL in H2O

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Molybdenum disulfide suspension (MoS2) is a 2D single layered transition metal dichalcogenide (TMD) in which hexagonal layers of molybdenum atoms are sandwiched between two layers of chalcogen atoms.[1]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

MoS2 can be used for a variety of applications such as ultrafast photonics, lithium ion batteries, and high speed flexible electronics.[1][2][3]
Owing to the thickness dependent physical and chemical properties, the transition metal dichalcogenide (TMD) based two dimensional (2D) nanomaterials have drawn significant attention recently.[4][5][6][7] The atomically thin layers of MoS2 and WS2 represent important class of TMD based 2D-material and they find applications in photocatalysis, phtotovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium ion battery applications of molybdenum disulfide (MoS 2) nanocomposites
Stephenson T, et al.
Energy & Environmental Science, 7(1), 209-231 (2014)
High-performance sensors based on molybdenum disulfide thin films
Lee K, et al.
Advanced Materials, 25(46), 6699-6702 (2013)
Molybdenum disulfide (MoS 2) as a broadband saturable absorber for ultra-fast photonics.
Zhang H, et al.
Optics Express, 22(6), 7249-7260 (2014)
Nikhita D Mansukhani et al.
Small (Weinheim an der Bergstrasse, Germany), 12(3), 294-300 (2015-12-01)
Conditions for the dispersion of molybdenum disulfide (MoS2) in aqueous solution at concentrations up to 0.12 mg mL(-1) using a range of nonionic, biocompatible block copolymers (i.e., Pluronics and Tetronics) are identified. Furthermore, the optimal Pluronic dispersant for MoS2 is
The super materials that could trump graphene.
Elizabeth Gibney
Nature, 522(7556), 274-276 (2015-06-19)

Articles

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Questions

  1. What is the melting point for the non-ionic surfactants in this product?

    1 answer
    1. The melting point range is 55-60°C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service