Skip to Content
Merck
All Photos(1)

Key Documents

808652

Sigma-Aldrich

Molybdenum disulfide

Crystal, 99.995%

Synonym(s):

2H-MoS2, Molybdenum (IV) sulfide, Molybdenum disulphide, Molybdenum sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,730.00

$1,730.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,730.00

About This Item

Empirical Formula (Hill Notation):
MoS2
CAS Number:
Molecular Weight:
160.07
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.23

$1,730.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.995%

form

crystals

L × W × H

0.5 cm × 0.5 cm × 0.1-0.15 mm, typical size, some variation in dimensions may occur with different batches

mp

2375 °C (lit.)

density

5.06 g/cm3 at 15 °C (lit.)

SMILES string

[Mo](=S)=S

InChI

1S/Mo.2S

InChI key

CWQXQMHSOZUFJS-UHFFFAOYSA-N

General description

Approximate dimensions of crystal: 0.5cm × 0.5cm × 0.1-0.15 mm. Some variation in size may occur.

Application

The two dimensional layered inorganic nanomaterials have drawn significant attention recently owing to their thickness dependent physical and chemical properties. They find applications in photocatalysis, phototovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc. Transition metal dischalcogenides represent an important class of inorganic 2D materials having the aforementioned applications.[1][2][3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Gunjakar JL, et al.
The Journal of Physical Chemistry C, 118, 3847-3863 (2014)
The super materials that could trump graphene.
Elizabeth Gibney
Nature, 522(7556), 274-276 (2015-06-19)
Sheneve Z Butler et al.
ACS nano, 7(4), 2898-2926 (2013-03-08)
Graphene's success has shown that it is possible to create stable, single and few-atom-thick layers of van der Waals materials, and also that these materials can exhibit fascinating and technologically useful properties. Here we review the state-of-the-art of 2D materials

Articles

Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.

Advances in scalable synthesis and processing of two-dimensional materials

For several decades, the need for an environmentally sustainable and commercially viable source of energy has driven extensive research aimed at achieving high efficiency power generation systems that can be manufactured at low cost.

Questions

  1. I am interested in your product 808652-1EA (crystal of MoS2), but I don't know what the size 1EA means... How big is this crystal (a x b x c) ? Can it be cut ? Is it a single crystal or a polycrystal ?

    1 answer
    1. This product is offered as a single crystal. The average dimensions are 0.5 cm (L) × 0.5 cm (W) × 0.1-0.15 mm (H) which is listed in the PROPERTIES section of the product page. The crystal is not sold by weight as the size will differ slightly from lot to lot. The expected net weight is 1 gram or less.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service