Skip to Content
Merck
All Photos(1)

Key Documents

901685

Sigma-Aldrich

Lithium hexafluorophosphate solution

in ethylene carbonate, dimethyl carbonate and diethyl carbonate, LiPF6 in EC/DMC/DEC=1:1:1 (v/v/v) 1.0 M, battery grade

Synonym(s):

1.0 M LiPF6 EC/DMC/DEC, 1M LiPF6 [EC/DMC/DEC=1:1:1 (v/v/v)], POWERLYTE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiPF6
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23
Pricing and availability is not currently available.

grade

battery grade

Quality Level

form

liquid

composition

LiPF6 in EC/DMC/DEC=1:1:1 (v/v/v), 1.0 M

concentration

(1.0 M LiPF6 in EC/DMC/DEC)

impurities

≤15 ppm H2O
≤50 ppm HF

anion traces

chloride (Cl-): ≤1 ppm
sulfate (SO42-): ≤2 ppm

cation traces

Ca: ≤1 ppm
Fe: ≤1 ppm
K: ≤1 ppm
Na: ≤1 ppm
Pb: ≤1 ppm

application(s)

battery manufacturing

SMILES string

F[P-](F)(F)(F)(F)F.[Li+]

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1

InChI key

AXPLOJNSKRXQPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium hexafluorophosphate solution in ethylene carbonate, dimethyl carbonate, and diethyl carbonate is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.[1][2][3][4]

Application

The solutions of LiPF6 in alkyl carbonates find applications as electrolytes in lithium ion batteries[5][6] and the use of high quality battery grade electrolytes having extremely low water (<15 PPM) and hydrogen fluoride (<50 PPM) contents are critical for achieving high electrochemical performance.

Caution

  • These electrolyte solutions have extremely low water content (<15ppm); Please handle under inert and mositure free enviornment (glove box).
  • Keep containers tightly closed. Keep away from heat and ignition sources.
  • Store in a cool and dry place. Avoid storing together with oxidizers.
  • Do not use any glass equipment.

Legal Information

Product of MU Ionic Solutions Corp.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 Oral

Target Organs

Bone,Teeth, Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

82.4 °F

Flash Point(C)

28 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Strategies for improving the cyclability and thermo-stability of LiMn2O4-based batteries at elevated temperatures.
Xu G, et al
Journal of Material Chemistry A, 3, 4092-4092 (2015)
Xu G, et al.
Journal of Material Chemistry A, 3, 4092-4092 (2015)
Lamb J, et al.
Journal of the Electrochemical Society, 162, A2131-A2131 (2015)
Electrodes with high power and high capacity for rechargeable lithium batteries
Kang K, et al.
Science, 311(5763), 977-980 (2006)
Challenges for rechargeable Li batteries
Goodenough JB and Kim Y
Chemistry of Materials, 22(3), 587-603 (2009)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service