Skip to Content
Merck
All Photos(1)

Key Documents

901661

Sigma-Aldrich

Zincate electroless immersion zinc plating solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$206.00

$206.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$206.00

About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

$206.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

color

colorless to light yellow

pH

>10

General description

Aluminum surfaces are not receptive to most plating processes due to their tendency to form oxides rapidly.Oxide formation inhibits uniform plating, resulting in inhomogeneous deposits, poor adhesion, or no deposition at all. By first applying Zincate Electroless Immersion Zinc Plating Solution to a properly prepared aluminum surface, it is possible subsequently deposit high quality electroless nickel films.

Application

Plating onto aluminum involves a multiple step process.
1.Non-etch clean the aluminum surface for up to 5 minutes with a detergent-based cleaner. Rinse thoroughly with cascading water.
2. De-smut the surface for 10-30 seconds using an acid mixture. RTM A for plastics is a suitable agent for smut removal. Rinse again thoroughly.
3. Mild etch for 5-10 seconds in a 25% solution of Aluminum Etch Type A. Rinse again thoroughly.
4. De-smut a second time and rinse as in step 2.
5. Apply the first Zincate layer. Immerse the part in the Zincate solution for 1-2 minutes at room temperature until a gray color develops. Rinse.
6. Remove the first Zincate layer using 50% nitric acid for 30-60 seconds or until the deposit turns white. Rinse.
7. Apply a second Zincate layer for 30-60 seconds. Rinse thoroughly before applying electroless nickel to the surface.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service