Skip to Content
Merck
All Photos(1)

Key Documents

901545

Sigma-Aldrich

Aluminum Etchant Type D

Aluminum Etchant Type D
1 of 1 reviewers received a sample product or took part in a promotion

Synonym(s):

Aluminum etching solution for use on gallium arsenide and gallium phosphide devices

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$97.80

$97.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$97.80

About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

$97.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

form

liquid

Related Categories

General description

ALUMINUM ETCHANT - TYPE D
  • Standard aluminum etchant for use on gallium arsenide, arsenide and gallium phosphide devices and for aluminum metallizations on nichrome resistors
  • Aluminum etchants are stable, non-toxic preparations used to etch aluminum metallizations on silicon devices and in integrated circuit applications. Aluminum contacts are defined, and interconnections are formed. These Aluminum Etchants, formulated with unique properties, easily overcome many of the difficulties experienced in aluminum etch processes.
  • The aluminum metallization and etching process using photo-lithographic techniques is basic to semiconductor and microelectronic technology. Aluminum etchants are highly compatible with commercial photoresists and permit delineation into high resolution patterns. Metal line width of 1 mil and separations less than 5 microns are feasible. High resolution is practical with these aluminum etchants because lifting of photoresist patterns does not occur and undercutting is minimized. Furthermore, the etchants do not attack silicon, silicon dioxide, silicon nitride or nichrome resistor films.
  • Two aluminum etchants are offered for use in microelectronics. Type A is recommended for use on silicon devices. Type D is recommended for use on gallium arsenide and gallium phosphide devices to avoid attack of the etchant on the intermetallic compound. It is also recommended for etching aluminum metallizations on nichrome thin film resistors.

Application

  • Aluminum metallizations up to 25,000 Å are vacuum deposited on the silicon slice, coated with a photoresist, and UV exposed using an appropriate photographic mask. The resist is developed to protect the aluminum where interconnections are desired. Then the unprotected areas of the aluminum are removed by etching with the aluminum etchant, followed by a water rinse.
  • Etching time is dependent upon the etchant temperature and the aluminum film thickness. When etching thick aluminum films, a higher etch rate is required; thus a higher etchant temperature should be used. Likewise, for thinner aluminum films, slower etch rates are desired and a lower etchant temperature should be chosen.
  • At a specific etchant temperature, the etching time is given by the following formula:
Etching time (second) + Film Thickness (Å)/ Etch Rate (Å/sec)
where Etch Rate at 25°C 30 Å/sec; at 40°C 80 Å/sec.

Features and Benefits

  • Controlled etch rates
  • Selective etching: will not attack SiO2 or Si3N4
  • Offers high resolution with minimal undercutting
  • Eliminates smut formation
  • Economical; reusable

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. London
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Great!

    Fast delivery, good etchant for GaAs-based semiconductors with aluminium surface layer

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service