Skip to Content
Merck
All Photos(1)

Key Documents

901307

Sigma-Aldrich

Polyester bis-MPA dendron

2 azide, 1 NHBoc (core), generation 1

Synonym(s):

Bis-MPA dendron, PFd-G1-BOCNH-Azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H57N7O13
Molecular Weight:
759.84
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

availability

available only in USA

color

white to off-white

storage temp.

−20°C

General description

  • Polymer architecture: Dendron
  • Generation: 1
  • Core functional group: Boc-protected amine
  • End Group Functionality: Azide
  • No. Surface Groups: 2
  • Calculated Mol. Wt.: 759.86 g/mol
  • Boc: tert-Butyloxycarbonyl
  • Bis-MPA: 2,2-Bis(hydroxymethyl)propionic acid

Application

Bis-MPA (or 2,2-Bis(methylol)propionic acid) is an aliphatic, pro-chiral molecule comprised of two hydroxyls and one carboxylic group that has been used in the synthesis of many different types of polymers. Dendrons synthesized from bis-MPA are biodegradable and have low cytotoxicity, making them well suited for use in biological research applications. These materials have been used in many applications such as signal amplification in bioassays and in drug delivery applications. The azide-functionalized dendrimers can be readily functionalized using either copper(I)-catalyzed alkyne-azide cycloaddition (CuAAC) or strain-promoted alkyne-azide cycloaddition (SPAAC). Additionally, the amine-functionalized core can be readily used in EDC or DCC coupling reactions (after Boc deprotection) with carbonyl-containing compounds to yield highly functionalized materials for a variety of biomedical applications.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service