Skip to Content
Merck
All Photos(1)

Key Documents

901091

Sigma-Aldrich

Zinc oxide ink for inkjet printing

greener alternative

Synonym(s):

Helios′Ink H-SZ01034 semiconductive ink, Zinc oxide dispersion, Zinc oxide suspension, ZnO ink, ZnO nanoparticle ink

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$341.00

$341.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$341.00

About This Item

Linear Formula:
ZnO
UNSPSC Code:
12352303
NACRES:
NA.23

$341.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Conductivity: 10-8-10-7 S/cm (before illumination) and 10-4-10-3 S/cm (under illumination with UV light)
Valence band: 7.9 ± 0.1 eV
Work function: 4.0 ± 0.1 eV

Quality Level

form

liquid (dispersion)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

slightly yellow to clear

surface tension

29 mN/m±5 mN/m

viscosity

10.5 mPa.s±3.0 mPa.s(20 °C)

capacity

≥0.9 % loading (Nanoparticles loading)

greener alternative category

General description

Helios′Ink H-SZ01034, semi conductive ink developed for the printed electronics is particularly well suited for OPV and PV. Helios′Ink H-SZ01034, semi conductive ink shows great performance on Drop on Demand (DOD) inkjet printers, blade coating, and spin coating and is compatible with various flexible substrates (polyimide, PET, PET/ITO, etc.) but also Glass/ITO.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Features and Benefits

Product benefits:
  • Easy deposition under atmospheric conditions (temperature and pressure).
  • Good optical performances (visible light transmission >90%).
  • Non CMR ink.
  • Compatible with ITO layer and Ag NWs layer.
  • Compatible with most flexible substrates.
  • Low drying temperature making printing onto flexible substrates possible.
  • Thin layers are obtained (20 nm) with low roughness (RMS = 3±1 nm).

Preparation Note

Typical processing guideline:
  • Can be homogenized for 5 minutes in an ultrasonic bath in order to get rid of any aggregates.
  • Filtration on a 0.45 μm PTFE filter syringe to avoid any nozzles clogging.
  • Drying conditions: Oven, IR oven, vacuum oven.
  • Clean-up solution: Ethanol/Acetone.

Legal Information

Product of Genes′Ink.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service