Please see this link: https://www.sigmaaldrich.com/sds/aldrich/901056.
901056
Carbon nanotube sheet
aligned, size × thickness 50 mm × 50 mm × 1-5 μm
Synonym(s):
MWCNT, MWNT, Multiwall carbon nanotube
Select a Size
About This Item
Recommended Products
Quality Level
Assay
>95% (carbon)
resistivity
10-1000 Ω/sq
size × thickness
50 mm × 50 mm × 1-5 μm
diameter
10-40 nm (carbon nanotubes)
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
CNT has a multi-walled structure. Crystal quality of CNT is examined by Raman scattering measurement. Intensity ratio of G-peak (1580 cm-1) and D-peak (1350 cm-1) represents quality of graphitization of carbon materials. G/D of CNT is ~3, showing high crystal quality. The high crystallinity is supported by transmission electron microscopy.
Preparation Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
CNT arrays: Vertically aligned nanotubes grown via CVD; drawn into pure, aligned sheets for innovations.
Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.
Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.
-
Hi Na Li, Do you have a SDS for the CNT sheet?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service