Skip to Content
Merck
All Photos(2)

Key Documents

724777

Sigma-Aldrich

Carbon nanotube, single-walled

≥90% carbon basis (≥80% as carbon nanotubes), 1-2 nm diameter, avg. no. of layers, 1

Synonym(s):

CNT, SWCNT, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$585.00
1 G
$1,760.00

$585.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$585.00
1 G
$1,760.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$585.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

G/D Ratio: ≥15 (Raman 633 nm)
Median length: 1 μm

Assay

≥90% carbon basis (≥80% as carbon nanotubes)

form

powder (freeze-dried)

feature

avg. no. of layers 1

manufacturer/tradename

Signis® CG200

surface area

≥700 m2/g

impurities

≤5 wt. % Moisture

diameter

1-2 nm

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

bulk density

0.1 g/cm3

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced using CHASM′s patented CoMoCAT synthesis technology, Signis® CG200 is the company′s largest-diameter single-wall carbon nanotube product.
Tube diameter determined by fluorescence spectroscopy.

Application

Carbon nanotube, single-walled (SWNT) belongs to the class of carbonaceous materials with excellent physiochemical, thermo-mechanical and electrochemical properties. This material can be used in a variety of sustainable energy applications such as solar cells, photocatalysis, thin film conductors, field effect transistors (FETs), biosensor, gas sensor, supercapacitor and nanomechanical resonators.[1][2][3][4][5][6][7]
Suitable for use in conductive composites and coatings.Functionalized single-walled carbon nanotubes (SWNTs) embedded in an insulating polymer matrix may be used as stable hole collection layer in perovskite solar cells.[8]

Preparation Note

CoMoCAT Catalytic Chemical Vapor Deposition (CVD) Method

Legal Information

CoMoCAT is a trademark of Chasm Advanced Materials
Signis is a registered trademark of Chasm Advanced Materials

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-walled carbon nanotubes increase
pandemic influenza A H1N1 virus infectivity of
lung epithelial cells
Sanpui P, et al.
Particle and Fibre Toxicology null
Sorting Carbon Nanotubes and Their Biological Applications
Oh H and Ju SY
Material Matters, 8(1) null
Carbon nanotube/polymer composites as a highly stable hole collection layer in perovskite solar cells.
Habisreutinger, Severin N., et al.
Nano Letters, 14(10), 5561-5568 (2014)
Single-walled carbon nanotubes as ultrahigh frequency nanomechanical resonators
Li C and Chou T
Physical Review. B, Condensed Matter and Materials Physics, 68(7), 073405-073405 (2003)
Charge transfer from carbon nanotubes to silicon in flexible carbon nanotube/silicon solar cells
Li X, et al.
Small, 13(48), 1702387-1702387 (2017)

Articles

The CoMoCAT® method of single-walled carbon nanotube (SWNT) synthesis yields high purity SWNTs with specific chiralities and narrow distributions of tube diameters.

Composite materials with micron scale reinforcements offer tailored properties for various applications.

Single-Walled Carbon Nanotubes synthesized by the Super-Growth Method & their properties & applications, including dispersing SGCNTs, SGCNT-polymer composites & SGCNT-metal composites are discussed.

Boron nitride nanotubes (BNNT) are close structural analogs of carbon nanotubes (CNT), which are high aspect ratio nanotubular material, where carbon atoms are alternately substituted by nitrogen and boron atoms.

See All

Protocols

Surfactant-assisted dispersion of single-walled carbon nanotubes for debundling or exfoliation in dispersion procedures.

Questions

1–3 of 3 Questions  
  1. Are these single-walled carbon nanotubes surface modified?

    1 answer
    1. No, these single-walled carbon nanotubes do not have surface modifications.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. How are these single-walled carbon nanotubes produced?

    1 answer
    1. These single-walled carbon nanotubes are produced by CoMoCAT® Catalytic Chemical Vapor Deposition (CVD).  Processing is done to remove amorphous carbon, and HF (aq) is used to remove the silica support and some metals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service