Skip to Content
Merck
All Photos(1)

Key Documents

901001

Sigma-Aldrich

4-Methoxycarbonylcubanecarboxylic acid

≥97%

Synonym(s):

(1s,2R,3R,4s,5s,6S,7S,8r)-4-(Methoxycarbonyl)cubane-1-carboxylic acid, Pentacyclo[4.2.0.02,5.03,8.04,7]octane-1,4-dicarboxylic acid, 1-methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$952.00

$952.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$952.00

About This Item

Empirical Formula (Hill Notation):
C11H10O4
CAS Number:
Molecular Weight:
206.19
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

$952.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

form

powder or crystals

mp

181 °C

functional group

carboxylic acid
ester

SMILES string

OC([C@]12[C@H]3[C@H]4[C@@H]1[C@H]5[C@@]4(C(OC)=O)[C@H]3[C@@H]52)=O

InChI

1S/C11H10O4/c1-15-9(14)11-5-2-6(11)4-7(11)3(5)10(2,4)8(12)13/h2-7H,1H3,(H,12,13)

InChI key

ARRJEFLYRAVFJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methoxycarbonylcubanecarboxylic acid is a tertiary carboxylic acid that can undergo iododecarboxylation using 1,3-diiodo-5,5-dimethylhydantoin (DIH) as both reaction initiator and iodine source under irradiative conditions. This product was previously listed as BML00136.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Metal-Free Efficient, General and Facile Iododecarboxylation Method with Biodegradable Co-Products.
Kulbitski K, et al.
Advanced Synthesis & Catalysis, 353(9), 1438-1442 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service