Skip to Content
Merck
4-Methoxycarbonylcubanecarboxylic acid ≥97%

4-Methoxycarbonylcubanecarboxylic acid

4-Methoxycarbonylcubanecarboxylic acid ≥97%
Empirical Formula (Hill Notation):
C11H10O4
CAS Number:
Molecular Weight:
206.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing