Skip to Content
Merck
All Photos(1)

Key Documents

900996

Sigma-Aldrich

2-(1H-Pyrrol-1-yl)ethanamine

Synonym(s):

1-(2-Aminoethyl)pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2
CAS Number:
Molecular Weight:
110.16
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

refractive index

n/D 1.5227

density

1.0136 g/mL

functional group

amine

SMILES string

NCCN1C=CC=C1

InChI

1S/C6H10N2/c7-3-6-8-4-1-2-5-8/h1-2,4-5H,3,6-7H2

InChI key

HPDIRFBJYSOVKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(1H-Pyrrol-1-yl)ethanamine may be used in the preparation of dihydropyrrolo[2′,1′:3,4]pyrazino[2,1-a]isoindolones by reacting with 2-formylbenzoic acids or 2-acetylbenzoic acid via N-acyliminium cation aromatic cyclizations. This product was previously listed as CBR00085.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

182.3 °F

Flash Point(C)

83.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convenient syntheses of dihydropyrrolo [2',1':3,4] pyrazino-and dihydropyrrolo [2',1':3,4][1,4] diazepino-[2,1-a] isoindolones.
Katritzky AR, et al.
Tetrahedron Letters, 43, 2831-2833 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service