Skip to Content
Merck
All Photos(2)

Key Documents

900965

Sigma-Aldrich

CzSi

Synonym(s):

9-(4-tert-Butylphenyl)-3,6-bis(triphenylsilyl)-9H-carbazole, 9-[4-(1,1-Dimethylethyl)phenyl]-3,6-bis(triphenylsilyl)-9H-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H49NSi2
CAS Number:
Molecular Weight:
816.19
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥97%

form

solid

InChI

1S/C58H49NSi2/c1-58(2,3)44-34-36-45(37-35-44)59-56-40-38-52(60(46-22-10-4-11-23-46,47-24-12-5-13-25-47)48-26-14-6-15-27-48)42-54(56)55-43-53(39-41-57(55)59)61(49-28-16-7-17-29-49,50-30-18-8-19-31-50)51-32-20-9-21-33-51/h4-43H,1-3H3

InChI key

WIHKEPSYODOQJR-UHFFFAOYSA-N

Application

Carbazole-based host materials for blue electrophosphorescence.
CzSi is a host material with tetrahedral Si atoms that act as effective spacers and thereby block the π conjugation of the carbazole core. It is used as a hole transporting molecule (HTM) that is mainly used in the development of organic light-emitting diodes (OLEDs). CzSi can also be used in the formation of a blue electrophosphorescence.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leakage-free solution-processed organic light-emitting diode using a ternary host with single-diode emission area up to 6times 11.5 cm 2
Liao S, et al.
Royal Society of Chemistry Advances, 9(19), 10584-10598 (2019)
Highly efficient blue-emitting iridium(III) carbene complexes and phosphorescent OLEDs.
Chiung-Fang Chang et al.
Angewandte Chemie (International ed. in English), 47(24), 4542-4545 (2008-05-08)
Highly efficient organic blue electrophosphorescent devices based on 3,6-bis(triphenylsilyl)carbazole as the host material.
Tsai MH, et al.
Advanced Materials, 18(9), 1216-1220 (2006)
Triphenylsilyl-and trityl-substituted carbazole-based host materials for blue electrophosphorescence
Tsai M, et al.
ACS Applied Materials & Interfaces, 1(3), 567-574 (2009)
Efficient blue organic light-emitting diodes employing thermally activated delayed fluorescence
Zhang et al.
Nature Photonics, 8(4), 326-332 (2014)

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service