Skip to Content
Merck
All Photos(1)

Key Documents

900926

Sigma-Aldrich

Branched PEI-g-PEG

PEG Mn 550

Synonym(s):

Branched polyethylenimine-graft-poly(ethylene glycol), PEI-g-PEG, PEI-PEG

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$333.00

$333.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$333.00

About This Item

Linear Formula:
H3CO[(CH2)2O]nC12H23N3O3[NH(CH2)2]mNH2
UNSPSC Code:
12162002
NACRES:
NA.23

$333.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

mol wt

PEG Mn 550

storage temp.

2-8°C

Application

Branched PEI-g-PEG, PEG Mn 550 was derived from Sigma-Aldrich Product 408727. Based upon an approximate Mw of 25,000 for the branched PEI, Product 900926 contains approximately 55 grafted PEG chains per PEI unit (by NMR).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Ladewig et al.
Journal of biomedical materials research. Part A, 102(7), 2137-2146 (2013-07-31)
High molecular weight (MW) polyethyleneimine (PEI) has been successfully used for the transfection of a broad variety of cell lines. In contrast to low MW PEI, which exhibits low transfection efficiencies but also low cytotoxicity, high MW PEI-mediated transfection achieves
Chunxi Liu et al.
Biomaterials, 34(10), 2547-2564 (2013-01-22)
Co-delivery of nucleic acids and chemotherapeutics has a potential to efficaciously treat human diseases via their synergetic effects. Activable therapeutic tools at the nanoscale are suitable platforms for combination therapy. In this study, we have developed a multifunctional nanoscaled delivery
Jing Wang et al.
Biomaterials science, 4(9), 1351-1360 (2016-07-19)
Zwitterionic poly(carboxybetaine) (PCB), poly(2-methacryloyloxyethyl phosphorylcholine) (PMPC) and non-ionic poly(ethylene glycol) (PEG), which have similar degrees of polymerization, were grafted to branched polyethyleneimine (PEI) to generate PCB-grafted PEI (PEI-PCB), PMPC-grafted PEI (PEI-PMPC) and PEG-grafted PEI (PEI-PEG) copolymers, respectively. These grafted PEI

Articles

Professor Yoshiki Katayama (Kyushu University, Japan) discusses recent advances in drug delivery systems and strategies that exploit the EPR effect, with a special focus on stimuli-responsive systems based on novel materials.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service