Skip to Content
Merck
All Photos(1)

Key Documents

767301

Sigma-Aldrich

Polyester bis-MPA dendron, 4 hydroxyl, 1 acetylene

generation 2, 95%

Synonym(s):

bisMPA dendrimer, bisMPA dendron

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$650.00

$650.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$650.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$650.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

hydroxyl surface groups

Assay

95%

form

solid

mol wt

generation 2

feature

focal point acetylene

no. Surface Groups

4

mp

101-105 °C

storage temp.

2-8°C

SMILES string

CC(CO)(CO)C(=O)OCC(C)(COC(=O)C(C)(CO)CO)C(=O)OCC#C

InChI

1S/C18H28O10/c1-5-6-26-15(25)18(4,11-27-13(23)16(2,7-19)8-20)12-28-14(24)17(3,9-21)10-22/h1,19-22H,6-12H2,2-4H3

InChI key

OENNSQAEPFFGAD-UHFFFAOYSA-N

General description

calculated mol. wt. 404.4

Application

Acetylene dendrons can be coupled to azide functional molecules such as carbohydrates, fluorescent dyes and polymers.

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methtyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service