Skip to Content
Merck
All Photos(1)

Key Documents

900803

Sigma-Aldrich

ITIC-DM

Synonym(s):

IT-2M

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$777.00

$777.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$777.00

About This Item

Empirical Formula (Hill Notation):
C98H90N4O2S4
CAS Number:
Molecular Weight:
1484.05
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

$777.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Band gap: 2.16 eV

Quality Level

Assay

99%

form

solid

solubility

organic solvents: soluble

Orbital energy

HOMO -5.45 eV 
LUMO -3.29 eV 

SMILES string

[s]1c2c([s]c(c2)\C=C%14/C(=O)c%15c(cc(c(c%15)C)C)C/%14=C(C#N)C#N)c3c1c4c(cc7c(c4)C(c%10c%11[s]c(cc%11[s]c%107)\C=C%12/C(=O)c%13c(cc(c(c%13)C)C)C/%12=C(C#N)C#N)(c9ccc(cc9)CCCCCC)c8ccc(cc8)CCCCCC)C3(c6ccc(cc6)CCCCCC)c5ccc(cc5)CCCCCC

InChI

1S/C98H90N4O2S4/c1-9-13-17-21-25-63-29-37-69(38-30-63)97(70-39-31-64(32-40-70)26-22-18-14-10-2)83-53-80-84(54-79(83)93-89(97)95-85(107-93)51-73(105-95)49-81-87(67(55-99)56-100)75-45-59(5)61(7)47-77(75)91(81)103)98(71-41-33-65(34-42-71)27-23-19-15-11-3,72-

InChI key

CJZPUZYWUPJTPK-UNAFEMLISA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunsun Li et al.
Advanced materials (Deerfield Beach, Fla.), 28(42), 9423-9429 (2016-09-09)
Fine energy-level modulations of small-molecule acceptors (SMAs) are realized via subtle chemical modifications on strong electron-withdrawing end-groups. The two new SMAs (IT-M and IT-DM) end-capped by methyl-modified dicycanovinylindan-1-one exhibit upshifted lowest unoccupied molecular orbital (LUMO) levels, and hence higher open-circuit

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service