Skip to Content
Merck
All Photos(1)

Documents

900400

Sigma-Aldrich

SLAP 3-SpiroCyHex N-Bn Pip Reagent

≥95%

Synonym(s):

1-((Benzyl((trimethylsilyl)methyl)amino)methyl)cyclohexanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32N2Si
Molecular Weight:
304.55
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

liquid

refractive index

n/D 1.516

density

0.956 g/mL

storage temp.

−20°C

InChI

1S/C18H32N2Si/c1-21(2,3)16-20(14-17-10-6-4-7-11-17)15-18(19)12-8-5-9-13-18/h4,6-7,10-11H,5,8-9,12-16,19H2,1-3H3

InChI key

HHDPYUUHZQUCNL-UHFFFAOYSA-N

Application

Silicon amine protocol (SLAP) reagents, in conjunction with aldehydes and ketones, form N-unprotected piperazines via photocatalytic cross-coupling with Ir[(ppy)2dtbbpy]PF6 (747769). Heteroaromatic, aromatic, and aliphatic aldehydes are well tolerated, as are structurally and stereochemically complex SLAP reagents. This product was introduced in collaboration with the Bode Research Group and provides a tin-free alternative to SnAP (tin amine protocol) reagents for piperazine synthesis.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Thiele et al.
ACS chemical biology, 7(12), 2004-2011 (2012-09-25)
Fatty acids are abundant constituents of all biological systems, and their metabolism is important for normal function at all levels of an organism. Aberrations in fatty acid metabolism are associated with pathological states and have become a focus of current
Keri A Tallman et al.
Journal of lipid research, 54(3), 859-868 (2013-01-12)
Monitoring lipid distribution and metabolism in cells and biological fluids poses many challenges because of the many molecular species and metabolic pathways that exist. This study describes the synthesis and study of molecules that contain an alkyne functional group as
Brent R Martin et al.
Nature methods, 6(2), 135-138 (2009-01-13)
S-palmitoylation is a pervasive post-translational modification required for the trafficking, compartmentalization and membrane tethering of many proteins. We demonstrate that the commercially available compound 17-octadecynoic acid (17-ODYA) can serve as a bioorthogonal, click chemistry probe for in situ labeling, identification
Xinxin Gao et al.
FEBS letters, 585(15), 2501-2506 (2011-07-12)
Palmitoylation of the Wnt and Hedgehog proteins is critical for maintaining their physiological functions. To date, there are no reported studies that characterize the cellular distribution of the palmitoylated forms of these proteins. Here, we describe the subcellular localization of
Stephen B Milne et al.
Nature chemical biology, 6(3), 205-207 (2010-01-26)
Alkyne-modified phospholipids can be unambiguously identified and differentiated from native species in complex mixtures by formation of dicobalthexacarbonyl complexes. This reaction is specific for alkynes and is unaffected by other glycerophospholipid-related moieties. Enrichment of cells with alkyne-derivatized fatty acids or

Protocols

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service