About This Item
Recommended Products
form
liquid
Quality Level
refractive index
n/D 1.489
density
1.122
functional group
ether
storage temp.
−20°C
SMILES string
CCCC[Sn](CCCC)(COCC1(CN(C(OC(C)(C)C)=O)CC1)N)CCCC
InChI
1S/C11H21N2O3.3C4H9.Sn/c1-10(2,3)16-9(14)13-6-5-11(12,7-13)8-15-4;3*1-3-4-2;/h4-8,12H2,1-3H3;3*1,3-4H2,2H3;
InChI key
JLZWEXDNLRNRBE-UHFFFAOYSA-N
Related Categories
General description
Application
Automate your N-heterocycle formation with Synple Automated Synthesis Platform (SYNPLE-SC002)
Other Notes
Professor product portal: Jeffrey Bode Research Group
SnAP Reagents for the Synthesis of Piperazines and Morpholines
SnAP reagents for the one-step synthesis of medium-ring saturated N-heterocycles from aldehydes
SnAP Reagents for a Cross-Coupling Approach to the One-Step Synthesis of Saturated N-Heterocycles
Bespoke SnAP Reagents for the Synthesis of C-Substituted Spirocyclic and Bicyclic Saturated N-Heterocycles
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
SnAP Reagents facilitate synthesis of saturated N-heterocycles for diverse structures.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service