Skip to Content
Merck
All Photos(1)

Documents

900381

Sigma-Aldrich

Uridine-15N2 5′-monophosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom % 15N, ≥95% (CP)

Synonym(s):

UMP-15N2 disodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H1115N2Na2O9P
Molecular Weight:
370.13
UNSPSC Code:
41106305
NACRES:
NA.12

isotopic purity

≥98 atom % 15N

Assay

≥95% (CP)

form

solution

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+2

storage temp.

−20°C

SMILES string

O=C(C=C[15N]1[C@@]2([H])[C@@](O)([H])[C@@](O)([H])[C@](COP(O[Na])(O[Na])=O)([H])O2)[15NH]C1=O

Related Categories

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service