900284
CyJohnPhos Pd G4
Synonym(s):
2-biphenylyl(dicyclohexyl)phosphine (Methanesulfonato-κO)[2′-(methylamino)-2-biphenylyl]palladium
Select a Size
Select a Size
About This Item
Recommended Products
form
powder
Quality Level
feature
generation 4
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings
functional group
phosphine
1 of 4
This Item | 900471 | 900349 | 900621 |
---|---|---|---|
form powder | form powder or crystals | form powder | form powder or crystals |
functional group phosphine | functional group phosphine | functional group phosphine | functional group phosphine |
feature generation 4 | feature generation 4 | feature generation 4 | feature generation 3 |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service