Merck
  • Highly efficient quantum-dot light-emitting diodes with DNA-CTMA as a combined hole-transporting and electron-blocking layer.
Highly efficient quantum-dot light-emitting diodes with DNA-CTMA as a combined hole-transporting and electron-blocking layer.
ACS nano (2009-03-25)
Qingjiang Sun, Guru Subramanyam, Liming Dai, Michael Check, Angela Campbell, Rajesh Naik, James Grote, Yongqiang Wang
ABSTRACT
Owing to their narrow bright emission band, broad size-tunable emission wavelength, superior photostability, and excellent flexible-substrate compatibility, light-emitting diodes based on quantum dots (QD-LEDs) are currently under intensive research and development for multiple consumer applications including flat-panel displays and flat lighting. However, their commercialization is still precluded by the slow development to date of efficient QD-LEDs as even the highest reported efficiency of 2.0% cannot favorably compete with their organic counterparts. Here, we report QD-LEDs with a record high efficiency (approximately 4%), high brightness (approximately 6580 cd/m(2)), low turn-on voltage (approximately 2.6 V), and significantly improved color purity by simply using deoxyribonucleic acid (DNA) complexed with cetyltrimetylammonium (CTMA) (DNA-CTMA) as a combined hole transporting and electron-blocking layer (HTL/EBL). This, together with controlled thermal decomposition of ligand molecules from the QD shell, represents a novel combined, but simple and very effective, approach toward the development of highly efficient QD-LEDs with a high color purity.
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, stabilized with octadecylamine ligands, fluorescence λem 620 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, stabilized with octadecylamine ligands, fluorescence λem 645 nm, solid
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 580 nm, 1 mg/mL in H2O
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 540 nm, 1 mg/mL in H2O
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, stabilized with octadecylamine ligands, fluorescence λem 520 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, stabilized with octadecylamine ligands, fluorescence λem 600 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 620 nm, 1 mg/mL in H2O