Skip to Content
Merck
All Photos(1)

Key Documents

900211

Sigma-Aldrich

Buchwald Precatalyst C-N Coupling Validation Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,650.00

$1,650.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$1,650.00

About This Item

UNSPSC Code:
12161600
NACRES:
NA.22

$1,650.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

Application

C-N Coupling Validation Kit allows users of theKITALYSIS-CN. to validate their hits- observed at 10 micromole scale- on larger scale, but can also be used as a stand-alone set of Pd catalysts. The following catalysts are included in the set:
Josiphos SL-J009-1 Pd G3 (747130-250MG)
SPhos Pd G2 (753009-250MG)
RuPhos Pd G2 (753246-250MG)
P(t-Bu)3 Pd G2 (756482-250MG)
BrettPhos Pd G3 (761605-250MG)
tBuXPhos Pd G3 (762229-250MG)
CPhos Pd G3 (763004-250MG)
XantPhos Pd G3 (763039-250MG)
XPhos Pd G3 (763381-250MG)
MorDalphos Pd G3 (792357-250MG)
DavePhos-Pd-G3 (804959-250MG)
rac-BINAP-Pd-G3 (804967-250MG)

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service