Skip to Content
Merck
All Photos(1)

Key Documents

722995

Sigma-Aldrich

4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid

Synonym(s):

4-Cyano-4-(thiobenzoylthio)pentanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$211.00
5 G
$692.00

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$211.00
5 G
$692.00

About This Item

Empirical Formula (Hill Notation):
C13H13NO2S2
CAS Number:
Molecular Weight:
279.38
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

94-98 °C

storage temp.

2-8°C

SMILES string

CC(CCC(O)=O)(SC(=S)c1ccccc1)C#N

InChI

1S/C13H13NO2S2/c1-13(9-14,8-7-11(15)16)18-12(17)10-5-3-2-4-6-10/h2-6H,7-8H2,1H3,(H,15,16)

InChI key

YNKQCPNHMVAWHN-UHFFFAOYSA-N

General description

4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid is asulfur-based chain transfer agent that provides a high degree of control forliving radical polymerization.

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of methacrylate and methacrylamide monomers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sieun Kim et al.
Biomacromolecules, 21(8), 3026-3037 (2020-07-17)
Charge anisotropy or the presence of charge patches at protein surfaces has long been thought to shift the coacervation curves of proteins and has been used to explain the ability of some proteins to coacervate on the "wrong side" of
Bartlomiej Kalaska et al.
Translational research : the journal of laboratory and clinical medicine, 177, 98-112 (2016-07-28)
The parenteral anticoagulants may cause uncontrolled and life-threatening bleeding. Protamine, the only registered heparin antidote, is partially effective against low-molecular weight heparins, completely ineffective against fondaparinux and may cause unacceptable toxicity. Therefore, we aimed to develop a synthetic compound for
Isadora Berlanga
Biomolecules, 9(8) (2019-08-11)
Giant vesicles with several-micrometer diameters were prepared by the self-assembly of an amphiphilic block copolymer in the presence of the Belousov-Zhabotinsky (BZ) reaction. The vesicle is composed of a non-uniform triblock copolymer synthesized by multi-step reactions in the presence of
Feng Liu et al.
Nanoscale, 8(7), 4387-4394 (2016-02-04)
The modulation of protein activity is of significance for disease therapy, molecular diagnostics, and tissue engineering. Nanoparticles offer a new platform for the preparation of protein conjugates with improved protein properties. In the present work, Escherichia coli (E. coli) inorganic
Hui-Yu Liu et al.
Molecules (Basel, Switzerland), 25(12) (2020-06-19)
Biomimetic lipid membranes on solid supports have been used in a plethora of applications, including as biosensors, in research on membrane proteins or as interfaces in cell experiments. For many of these applications, structured lipid membranes, e.g., in the form

Articles

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

RAFT polymerization offers living characteristics to radical polymerization, contributing versatility to reversible deactivation radical polymerization methods.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

See All

Protocols

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service