Skip to Content
Merck
All Photos(1)

Documents

88753

Sigma-Aldrich

(R)-1-[(SP)-2-(Di-tert-butylphosphino)ferrocenyl]ethyldiphenylphosphine

≥97%

Synonym(s):

(1R)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene (acc to CAS), Josiphos SL-J502-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40FeP2
CAS Number:
Molecular Weight:
542.45
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97%

form

powder

optical purity

ee: ≥99%

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(C(C)(C)C)C(C)(C)C)P(c3ccccc3)c4ccccc4

InChI

1S/C27H35P2.C5H5.Fe/c1-21(24-19-14-20-25(24)29(26(2,3)4)27(5,6)7)28(22-15-10-8-11-16-22)23-17-12-9-13-18-23;1-2-4-5-3-1;/h8-21H,1-7H3;1-5H;/t21-;;/m1../s1

InChI key

MJOXBJGLXGTJRJ-GHVWMZMZSA-N

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service