Skip to Content
Merck
All Photos(2)

Key Documents

259098

Sigma-Aldrich

(2S,3S)-(−)-Bis(diphenylphosphino)butane

Synonym(s):

(2S,3S)-(−)-2,3-Bis(diphenylphosphino)butane, (S,S)-Chiraphos

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$253.00
1 G
$955.00

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$253.00
1 G
$955.00

About This Item

Linear Formula:
[-CH(CH3)P(C6H5)2]2
CAS Number:
Molecular Weight:
426.47
Beilstein:
4268869
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

optical activity

[α]22/D −191°, c = 1.5 in chloroform

mp

108-110 °C (lit.)

functional group

phosphine

SMILES string

C[C@@H]([C@H](C)P(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C28H28P2/c1-23(29(25-15-7-3-8-16-25)26-17-9-4-10-18-26)24(2)30(27-19-11-5-12-20-27)28-21-13-6-14-22-28/h3-24H,1-2H3/t23-,24-/m0/s1

InChI key

FWXAUDSWDBGCMN-ZEQRLZLVSA-N

Application

Ligand for Pd-catalyzed carbon-carbon bond formation.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moinet, C. Fiaud, J.-C.
Tetrahedron Letters, 36, 2051-2051 (1995)
M. Yamaguchi et al.
Tetrahedron Asymmetry, 2, 663-663 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service