Skip to Content
Merck
All Photos(1)

Key Documents

88128

Sigma-Aldrich

Tetrapropylammonium tetrafluoroborate

≥98.0%

Synonym(s):

Tetrapropylammonium tetrafluoroborate(1-), Tetrapropylammoniumtetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$340.00

$340.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$340.00

About This Item

Linear Formula:
(CH3CH2CH2)4N(BF4)
CAS Number:
Molecular Weight:
273.16
Beilstein:
3637590
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$340.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0%

form

crystals

mp

245-247 °C

SMILES string

F[B-](F)(F)F.CCC[N+](CCC)(CCC)CCC

InChI

1S/C12H28N.BF4/c1-5-9-13(10-6-2,11-7-3)12-8-4;2-1(3,4)5/h5-12H2,1-4H3;/q+1;-1

InChI key

ADADJCUTHQJPCB-UHFFFAOYSA-N

Application

Tetrapropylammonium tetrafluoroborate (Pr4NBF4 or TPABF4) can be used as:
  • An electrolyte in the determination of phenolic compounds from grape seed extracts by capillary electrophoretic method.[1]
  • A supporting electrolyte in the electrochemical hydrogenation of dimethylformamide (DMF) to trimethylamine (TMA).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical synthesis of trimethylamine by cathodic reduction of dimethylformamide using Cu and Pd cathodes
Jia R, et al.
Journal of Electroanalytical Chemistry, 741(11), 32-35 (2015)
Tetraethylammonium tetrafluoroborate: a novel electrolyte with a unique role in the capillary electrophoretic separation of polyphenols found in grape seed extracts
Yanes EG, et al.
Analyst, 125(11), 1919-1923 (2000)
Ludmila Šimková et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 323, 106895-106895 (2021-01-12)
The molecule of 2,2-dinitroethene-1,1-diamine (FOX-7) is one of the most interesting molecules with multiple redox centres stabilized by push-pull effect. To reveal the detailed mechanism of its electrochemical process radical intermediates formed in the course of its electrochemical reduction in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service